Learn More
Electron Microscopy Sciences Crossed Scale Microscope Graticle NE 72, Custom, 20 mm with 200 divisions of 0.1 mm
SDP
Catalog No. 501901779
Encompass_Preferred
Used as a horizontal and a vertical scale, they are especially useful when interested in measurement in different axis. NE 72 Crossed Micrometer scale.
Size: Custom
Divisions: 20 mm with 200 divisions of 0.1 mm
Catalog No. 50-190-1779 Supplier Electron Microscopy Sciences Supplier No. 6797850
Used as a horizontal and a vertical scale, they are especially useful when interested in measurement in different axis. NE 72 Crossed Micrometer scale.
Size: Custom
Divisions: 20 mm with 200 divisions of 0.1 mm