Learn More
Electron Microscopy Sciences Scale with Cross Line Microscope Graticle NE777, Custom, 0.5" with divisions of 0.005"
SDP
Catalog No. 501901788
Encompass_Preferred
Used as a horizontal and a vertical scale, they are especially useful when interested in measurement in different axis. Horizontal micrometer scale 0.5" long with divisions of 0.005".
Size: Custom
Divisions: 0.5" with divisions of 0.005"
Catalog No. 50-190-1788 Supplier Electron Microscopy Sciences Supplier No. 6798050
Used as a horizontal and a vertical scale, they are especially useful when interested in measurement in different axis. Horizontal micrometer scale 0.5" long with divisions of 0.005".
Size: Custom
Divisions: 0.5" with divisions of 0.005"