Learn More
Electron Microscopy Sciences Cross Lines Graticle NE8, 23 mm, Nominal line width 0.020 mm
SDP
Catalog No. 501927976
Encompass_Preferred
Used as NE50 but for measurements in two directions and for sighting and alignment. Image covers the entire field of view.
Size: 23 mm
Divisions: Nominal line width 0.020 mm
Surface: Chrome image
Catalog No. 50-192-7976 Supplier Electron Microscopy Sciences Supplier No. 6798323
Used as NE50 but for measurements in two directions and for sighting and alignment. Image covers the entire field of view.
Size: 23 mm
Divisions: Nominal line width 0.020 mm
Surface: Chrome image