Learn More
Electron Microscopy Sciences Lennox Grain Analysis Graticle NG21, 16mm
SDP
Catalog No. 501901804
Encompass_Preferred
The eyepiece reticle is used for Lennox grain size analysis. The pattern comprises and array of dashed squares and crosses.
Size: 16 mm
Catalog No. 50-190-1804 Supplier Electron Microscopy Sciences Supplier No. 6800416
The eyepiece reticle is used for Lennox grain size analysis. The pattern comprises and array of dashed squares and crosses.
Size: 16 mm