Learn More
Electron Microscopy Sciences Lennox Grain Analysis Graticle NG21, 21mm
SDP
Catalog No. 501901806
Encompass_Preferred
The eyepiece reticle is used for Lennox grain size analysis. The pattern comprises and array of dashed squares and crosses.
Size: 21 mm
Catalog No. 50-190-1806 Supplier Electron Microscopy Sciences Supplier No. 6800421
The eyepiece reticle is used for Lennox grain size analysis. The pattern comprises and array of dashed squares and crosses.
Size: 21 mm