Learn More
Electron Microscopy Sciences Lennox Grain Analysis Graticle NG21, Custom
SDP
Catalog No. 501902075
Encompass_Preferred
The eyepiece reticle is used for Lennox grain size analysis. The pattern comprises and array of dashed squares and crosses.
Size: Custom
Catalog No. 50-190-2075 Supplier Electron Microscopy Sciences Supplier No. 6800450
The eyepiece reticle is used for Lennox grain size analysis. The pattern comprises and array of dashed squares and crosses.
Size: Custom