Learn More
Electron Microscopy Sciences Grain Sizing Reticle NG60, 21 mm
SDP
Catalog No. 501901808
Encompass_Preferred
For the determination of non-metallic inclusion content of steel. NG60 meets EN 10247 and NG61 Meets ISO 4976. Both are scaled for use eith 10x objective magnification Pattern
Size: 21 mm
Method: EN 10247
Catalog No. 50-190-1808 Supplier Electron Microscopy Sciences Supplier No. 6800721
For the determination of non-metallic inclusion content of steel. NG60 meets EN 10247 and NG61 Meets ISO 4976. Both are scaled for use eith 10x objective magnification Pattern
Size: 21 mm
Method: EN 10247