Learn More
Electron Microscopy Sciences Grain Sizing Reticle NG61, 24.5 mm
SDP
Catalog No. 501901815
Encompass_Preferred
For the determination of non-metallic inclusion content of steel. NG60 meets EN 10247 and NG61 Meets ISO 4976. Both are scaled for use eith 10x objective magnification Pattern
Size: 24.5 mm
Method: ISO 4967
Catalog No. 50-190-1815 Supplier Electron Microscopy Sciences Supplier No. 6803624
For the determination of non-metallic inclusion content of steel. NG60 meets EN 10247 and NG61 Meets ISO 4976. Both are scaled for use eith 10x objective magnification Pattern
Size: 24.5 mm
Method: ISO 4967