Learn More
Electron Microscopy Sciences Grating Replica, Crossed Line, 1 each
SDP
Catalog No. 5026393
Encompass_Preferred
Crossed: Cross line pitch spacing of 463nm with 2160 lines/mm in both directions, cross at 90 to one another give additional accuracy to magnification checks and aid in checking distortion. Trench-type groove makes it easy to measure pitch. Ideal for calibration electron-optical magnification up to the x80000/100000 range.
Catalog No. 50-263-93 Supplier Electron Microscopy Sciences Supplier No. 80051
Crossed: Cross line pitch spacing of 463nm with 2160 lines/mm in both directions, cross at 90 to one another give additional accuracy to magnification checks and aid in checking distortion. Trench-type groove makes it easy to measure pitch. Ideal for calibration electron-optical magnification up to the x80000/100000 range.