Learn More
Electron Microscopy Sciences Grating Replica On Latex Spheres
SDP
Catalog No. 5026951
Encompass_Preferred
The latex sphere is 0.261 microns and the grating replica is 2160 lines/mm. This calibration id allows you to double check the accuracy of magnification calibration.
Catalog No. 50-269-51 Supplier Electron Microscopy Sciences Supplier No. 80055
The latex sphere is 0.261 microns and the grating replica is 2160 lines/mm. This calibration id allows you to double check the accuracy of magnification calibration.