Learn More
Electron Microscopy Sciences Grating Replica Parallel Line
SDP
Catalog No. 5026392
Encompass_Preferred
Shadowcast carbon replica of diffraction parallel line gratings with spacing of 462.9 nm, 2160 lines per millimeter onto 3.05mm grid.
Catalog No. 50-263-92 Supplier Electron Microscopy Sciences Supplier No. 80050
Shadowcast carbon replica of diffraction parallel line gratings with spacing of 462.9 nm, 2160 lines per millimeter onto 3.05mm grid.