Learn More
Electron Microscopy Sciences Optical Resolution Grating with Ronchi Ruling 200 lines/ mm
SDP
Catalog No. 501901825
Encompass_Preferred
Metallic grating pattern on glass squares with opaque lines approximately 50% of the pitch. Stock items are on 50 mm x 50 mm squares of glass 1.5 mm thick. The glass is B270 with refractive index of 1.52. Mutually perpendicular pairs produce a screen of square apertures. Vacuum deposited chrome gratings on glass substrate. Typically used for testing of resolution, field distortion and parfocal stability, but may be paired to provide interference patterns. 26mm x 10mm soda. 200 lines per mm ruled over 30mm
Catalog No. 50-190-1825 Supplier Electron Microscopy Sciences Supplier No. 6809832
Metallic grating pattern on glass squares with opaque lines approximately 50% of the pitch. Stock items are on 50 mm x 50 mm squares of glass 1.5 mm thick. The glass is B270 with refractive index of 1.52. Mutually perpendicular pairs produce a screen of square apertures. Vacuum deposited chrome gratings on glass substrate. Typically used for testing of resolution, field distortion and parfocal stability, but may be paired to provide interference patterns. 26mm x 10mm soda. 200 lines per mm ruled over 30mm