Learn More
Electron Microscopy Sciences Optical Resolution Grating with Ronchi Ruling 40 lines/ mm
SDP
Catalog No. 501901589
Encompass_Preferred
Metallic grating pattern on glass squares with opaque lines approximately 50% of the pitch. Stock items are on 50 mm x 50 mm squares of glass 1.5 mm thick. The glass is B270 with refractive index of 1.52. Mutually perpendicular pairs produce a screen of square apertures. Vacuum deposited chrome gratings on glass substrate. Typically used for testing of resolution, field distortion and parfocal stability, but may be paired to provide interference patterns. 50mm x 50mm B270 glass. 40 lines per mm ruled over full area
Catalog No. 50-190-1589 Supplier Electron Microscopy Sciences Supplier No. 6809828
Metallic grating pattern on glass squares with opaque lines approximately 50% of the pitch. Stock items are on 50 mm x 50 mm squares of glass 1.5 mm thick. The glass is B270 with refractive index of 1.52. Mutually perpendicular pairs produce a screen of square apertures. Vacuum deposited chrome gratings on glass substrate. Typically used for testing of resolution, field distortion and parfocal stability, but may be paired to provide interference patterns. 50mm x 50mm B270 glass. 40 lines per mm ruled over full area