Learn More
Surgical Design™ Graves Vaginal Speculum
Catalog No. 21125067 Shop All Surgical Design Products
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
21-125-067 Large
21-125-065 Small
21-125-066 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 21-125-067 Supplier Surgical Design™ Supplier No. ORG275
Add to Cart
Add to Cart
Graves vaginal speculum provides durable, smooth-action performance for reliable visualization during gynecological examinations.
  • Available Sizes: Small, Medium, Large
  • Traditional bivalve design provides wide, adjustable visualization
  • Smooth screw mechanism ensures controlled and stable opening
  • Rounded, polished edges enhance patient comfort
  • Medical-grade stainless steel—autoclavable and corrosion-resistant
  • Suitable for OB/GYN clinics, emergency medicine, general exams, and procedural setups
Specifications
Product Type Vaginal Speculum
Grade Medical
Size Large
Autoclavable Autoclavable