Learn More
Mitegen GraVitri Basic Manual Plunger
SDP
Catalog No. 5000494990
Encompass_Preferred
Designed for ultimate accessibility and affordability, the GraVitri Basic and Standard units simplify the plunge-freezing workflow and reduce overhead. Since they require no power or external connections, setup is instant, maintenance is minimal, and operational costs are significantly lower. The system is modular for straightforward expansion as your science grows.
Catalog No. 50-004-94990 Supplier Mitegen Supplier No. MGPC1B
Designed for ultimate accessibility and affordability, the GraVitri Basic and Standard units simplify the plunge-freezing workflow and reduce overhead. Since they require no power or external connections, setup is instant, maintenance is minimal, and operational costs are significantly lower. The system is modular for straightforward expansion as your science grows.