Learn More
Thermco Products Inc ASTM Approved Applications, Blue Spirit, ASTM S12F, Gravity, -5 deg./215 deg.F, 0.5 deg.F div, TOTAL imm, 425mm
Supplier:  Thermco Products Inc ACC012FBLS
Encompass_Preferred
The ASTM S12F is made in accordance with specifications of the American Society for Testing and Material. Strict manufacturing procedures and quality controls insure full compliance to ASTM E-1. This instrument are made from annealed glass and have been extensively aged for minimal scale error and stability. The ASTM S12F is used in Gravity proceedures in the petroleum industry.
Catalog No. NC0554874