Learn More
MARVELGENT BIOSCIENCES INC Empty Disposable Gravity Flow Columns, 300 mL with 50 um frits, 10/pk
SDP
Catalog No. NC1804313
Encompass_Preferred
These disposable columns are economical, versatile columns for single-use purification applications using gravity flow. Simple, cost-effective, readily adaptable for a wide range of gravity flow applications. Made of high purity medical grade polypropylene, stable in commonly used chemicals and reagents. Available in various sizes to suit for different experiments.
Catalog No. NC1804313 Supplier MARVELGENT BIOSCIENCES INC Supplier No. 120280010
These disposable columns are economical, versatile columns for single-use purification applications using gravity flow. Simple, cost-effective, readily adaptable for a wide range of gravity flow applications. Made of high purity medical grade polypropylene, stable in commonly used chemicals and reagents. Available in various sizes to suit for different experiments.