Learn More
Med Vet International Gray Cold Sterilization Tray, 10" x 5" x 3", J0882B
Catalog No. 5000584992
Encompass_Preferred
Durable plastic made to withstand disinfectants and sterilants. All have drain baskets that allow lifting the tray out of solution and having the solution drain back into the main container. Equipped with fitted sealed lid. Soaking Tray with hinged lid. Handled drain basket. 5"W x 101/2"L x 31/4"H.
Catalog No. 50-005-84992 Supplier Med Vet International Supplier No. JOR882B
Durable plastic made to withstand disinfectants and sterilants. All have drain baskets that allow lifting the tray out of solution and having the solution drain back into the main container. Equipped with fitted sealed lid. Soaking Tray with hinged lid. Handled drain basket. 5"W x 101/2"L x 31/4"H.