Learn More
Sklar Instruments Gray Cystic Duct Forceps, Sklar, Serrated with 1x2 Teeth, Long Curve, 9 in., Stainless Steel. EA/1.
SDP
Catalog No. 501438205 Shop All Sklar Instruments Products
Encompass_Preferred
Gray Cystic Duct Forceps, Sklar, Serrated with 1x2 Teeth, Long Curve, 9 in., Stainless Steel. Sklar's Gray cystic duct forceps are used in gall bladder procedures, such as cholecystectomy. They ar ratcheted finger ring instruments with a serrated curved tip for effective gripping. They come in a variety of lengths, curvatured, and toothed or non-toothed.
This item is not returnable. View return policy
Catalog No. 50-143-8205 Supplier Sklar Instruments Supplier No. 322095
Gray Cystic Duct Forceps, Sklar, Serrated with 1x2 Teeth, Long Curve, 9 in., Stainless Steel. Sklar's Gray cystic duct forceps are used in gall bladder procedures, such as cholecystectomy. They ar ratcheted finger ring instruments with a serrated curved tip for effective gripping. They come in a variety of lengths, curvatured, and toothed or non-toothed.