Learn More
EZKEM Pump Tubing Gray/Gray 0.051 ID 6x36 12pk
SDP
Catalog No. NC1547968
Encompass_Preferred
EZkem’s 2-stop 6″ collar x 36″ length peristaltic pump tubing is used for Ismatec peristaltic pumps in Alpkem, OI Analytical, Latchat and other brands of autoanalyzers for flow injection (FIA) and segmented flow (SFA) analysis. A000352 Gray/Gray 0.051 ID 12/pk
Catalog No. NC1547968 Supplier EZKEM Supplier No. A000352
EZkem’s 2-stop 6″ collar x 36″ length peristaltic pump tubing is used for Ismatec peristaltic pumps in Alpkem, OI Analytical, Latchat and other brands of autoanalyzers for flow injection (FIA) and segmented flow (SFA) analysis. A000352 Gray/Gray 0.051 ID 12/pk