Learn More
Agilent Technologies Tubing, PVC, for aqueous samples, gray/gray, 12/pk
Catalog No. NC1777923
Encompass
Suitable for all aqueous solutions, medium-concentrated acids and bases.
Catalog No. NC1777923 Supplier Agilent Technologies Supplier No. 3710034500
Suitable for all aqueous solutions, medium-concentrated acids and bases.