Learn More
Selleck Chemical LLC Grazoprevir 2mg 1350514-68-9 MK5172
SDP
Catalog No. 501948292
Encompass_Preferred
Grazoprevir anhydrous (MK5172) is a Hepatitis C Virus NS3/4A Protease inhibitor with IC50 values of 7pM, 4pM, and 62pM for HCV genotype 1a, 1B, and 4 respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8292 Supplier Selleck Chemical LLC Supplier No. S37282MG
Grazoprevir anhydrous (MK5172) is a Hepatitis C Virus NS3/4A Protease inhibitor with IC50 values of 7pM, 4pM, and 62pM for HCV genotype 1a, 1B, and 4 respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.