Learn More
Medchemexpress LLC Grazoprevir potassium salt | 1206524-86-8 | C38H49KN6O9S | 1 ML
SDP
Catalog No. 5000373618
Encompass_Preferred
Grazoprevir potassium salt is a selective inhibitor of Hepatitis C virus NS3/4a protease with broad activity across various genotypes and resistant variants. It also inhibits SARS-CoV-2 3CLpro activity. This product is for research use only.
  • Selective inhibitor of Hepatitis C virus NS3/4a protease
  • Broad activity across various genotypes and resistant variants
  • Inhibits SARS-CoV-2 3CLpro activity
  • Potent against HCV replication mutants NS5A (Y93H), NS5B nucleosides (S282T), and NS5B (C316Y)
  • Demonstrates in vivo efficacy against chronic-HCV-infected chimpanzees
Catalog No. 50-003-73618 Supplier Medchemexpress LLC Supplier No. HY15298A10MM/1ML
Grazoprevir potassium salt is a selective inhibitor of Hepatitis C virus NS3/4a protease with broad activity across various genotypes and resistant variants. It also inhibits SARS-CoV-2 3CLpro activity. This product is for research use only.
  • Selective inhibitor of Hepatitis C virus NS3/4a protease
  • Broad activity across various genotypes and resistant variants
  • Inhibits SARS-CoV-2 3CLpro activity
  • Potent against HCV replication mutants NS5A (Y93H), NS5B nucleosides (S282T), and NS5B (C316Y)
  • Demonstrates in vivo efficacy against chronic-HCV-infected chimpanzees