Learn More
Medchemexpress LLC Grazoprevir potassium salt | 1206524-86-8 | C38H49KN6O9S | 50 MG
SDP
Catalog No. 5000373180
Encompass_Preferred
Grazoprevir potassium salt is a selective inhibitor of Hepatitis C virus (HCV) NS3/4a protease, demonstrating broad activity across various genotypes and resistant variants. It also inhibits SARS-CoV-2 3CLpro activity. This compound has shown efficacy in vivo against chronic-HCV-infected chimpanzees and good plasma and liver exposure in dogs, highlighting its potential as an antiviral agent.
  • Selective inhibitor of Hepatitis C virus NS3/4a protease
  • Broad activity across genotypes and resistant variants
  • Effective against major genotypes and common resistant mutations
  • Inhibits SARS-CoV-2 3CLpro activity
  • Demonstrates subnanomolar to low-nanomolar EC50s against genotypes 1a, 1b, and 2a
  • Maintains excellent potency against HCV replication mutants
  • Shows efficacy against chronic-HCV-infected chimpanzees
  • Achieves good plasma and high liver exposure in dogs
Catalog No. 50-003-73180 Supplier Medchemexpress LLC Supplier No. HY15298A50MG
Add to Cart
Add to Cart
Grazoprevir potassium salt is a selective inhibitor of Hepatitis C virus (HCV) NS3/4a protease, demonstrating broad activity across various genotypes and resistant variants. It also inhibits SARS-CoV-2 3CLpro activity. This compound has shown efficacy in vivo against chronic-HCV-infected chimpanzees and good plasma and liver exposure in dogs, highlighting its potential as an antiviral agent.
  • Selective inhibitor of Hepatitis C virus NS3/4a protease
  • Broad activity across genotypes and resistant variants
  • Effective against major genotypes and common resistant mutations
  • Inhibits SARS-CoV-2 3CLpro activity
  • Demonstrates subnanomolar to low-nanomolar EC50s against genotypes 1a, 1b, and 2a
  • Maintains excellent potency against HCV replication mutants
  • Shows efficacy against chronic-HCV-infected chimpanzees
  • Achieves good plasma and high liver exposure in dogs