Learn More
Aviva Systems Biology Corp GRB2 Antibody - N-terminal region (ARP58474_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58474_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GRB2. It was validated on Western Blot. . Alternate names: ASH, EGFRBP-GRB2, Grb3-3, MST084, MSTP084, NCKAP2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-479