Learn More
Sino Biological Recombinant Human WWP1 Protein (Full Length, GST Tag)
Catalog No. 502572553
Encompass_Preferred
Recombinant full length human WWP1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-2553 Supplier Sino Biological Supplier No. W29630G100
Recombinant full length human WWP1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.