Learn More
Grainger Westward Bulk Loader Valve, 0.34 in. Diameter, 1/8 in. MNPT, 1 ea
Catalog No. 502756550
Encompass_Preferred
The bulk loader valve is designed for the efficient bulk filling of grease guns. This valve facilitates clean and rapid grease transfer, ensuring compatibility with standard grease filler pumps equipped with straight loader adaptors.
  • Fits grease filler pumps with straight loader adaptors such as those from Lincoln
  • Features a 0.34 in. overall diameter
  • Includes a ball check mechanism
  • Equipped with a 1/8 in. MNPT fitting end
  • Straight head angle design
Catalog No. 50-275-6550 Supplier Grainger Supplier No. 15F206
The bulk loader valve is designed for the efficient bulk filling of grease guns. This valve facilitates clean and rapid grease transfer, ensuring compatibility with standard grease filler pumps equipped with straight loader adaptors.
  • Fits grease filler pumps with straight loader adaptors such as those from Lincoln
  • Features a 0.34 in. overall diameter
  • Includes a ball check mechanism
  • Equipped with a 1/8 in. MNPT fitting end
  • Straight head angle design