Learn More
Grainger Westward Grease Fitting Thread Tester, Aluminum, 21 Holes, 1 ea
Catalog No. 502770777
Encompass_Preferred
This grease fitting thread tester is a precision tool designed to accurately identify thread types and sizes for grease fittings. Featuring an anodized aluminum construction, this gage provides durability and readability for both inch and metric measurements.
  • Supports multiple thread types including BSF, BSPT, BSW, Metric, NPT, and UNF
  • Constructed from anodized aluminum for durability
  • Contains 21 holes to accommodate various fitting sizes
  • Measures diameters ranging from 0.125 in. to 0.375 in.
  • Dimensions: 8 3/16 in. length by 3 in. width by 15/64 in. thickness
Catalog No. 50-277-0777 Supplier Grainger Supplier No. 13X066
This grease fitting thread tester is a precision tool designed to accurately identify thread types and sizes for grease fittings. Featuring an anodized aluminum construction, this gage provides durability and readability for both inch and metric measurements.
  • Supports multiple thread types including BSF, BSPT, BSW, Metric, NPT, and UNF
  • Constructed from anodized aluminum for durability
  • Contains 21 holes to accommodate various fitting sizes
  • Measures diameters ranging from 0.125 in. to 0.375 in.
  • Dimensions: 8 3/16 in. length by 3 in. width by 15/64 in. thickness