Learn More
Synthware™ Grease-Free, Two-Piece Sublimator
GREENER_CHOICESDP
Catalog No. 31502084 Shop All Kemtech Products
Change view
Click to view available options
Capacity (Metric):
10 mL
40 mL
60 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
31-502-084 60 mL
31-502-083 40 mL
31-502-082 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-502-084 Supplier Synthware™ Supplier No. S735060
Add to Cart
Add to Cart
A grease-free, two-piece sublimation apparatus consisting of an upper water cooled condenser and lower vessel to hold the material to be sublimed.
  • The sublimate collects on the lower end of the condenser which has a flat bottom
  • Condenser is constructed using a grease-free, threaded valve
  • Connection of the condenser and lower vessel via an O-ring joint enables the two parts to be gently separated for maximum product recovery
  • Complete apparatus consists of the cold finger condenser, lower vessel, Viton™ O-ring and stainless steel pinch clamp
Specifications
For Use With (Application) Organic Synthesis
Material Borosilicate Glass
Quantity 1 Ea.
Capacity (Metric) 60 mL
Product Type Water Cooled