Learn More
Netmercury GREASE KRYTOX GPL205 2OZ
Catalog No. NC9534759
Encompass
GREASE KRYTOX GPL205 2OZ - Krytox GPL 200-207 greases are white buttery greases with all of the same properties as the Krytox GPL 100-107 oils that are made from, but they are in grease form
Catalog No. NC9534759 Supplier Netmercury Supplier No. NM00020135
GREASE KRYTOX GPL205 2OZ - Krytox GPL 200-207 greases are white buttery greases with all of the same properties as the Krytox GPL 100-107 oils that are made from, but they are in grease form