Learn More
Grainger Westward Grease Pump, Nitrile and Steel, 8,700 psi, 1 ea
Catalog No. 502755040
Encompass_Preferred
This high-pressure, foot-operated portable grease pump is designed for efficient, effortless lubrication of automotive, agricultural, and earth-moving equipment. The system features a design where all moving parts remain submerged in grease, which creates a wave action during operation to automatically eliminate trapped air pockets and ensure continuous lubrication without requiring a follower plate.
  • Delivers 0.07 oz. per stroke at a maximum pressure of 8,700 psi.
  • Compatible with NLGI 1 and 2 grease.
  • Operated via a convenient foot lever for high-pressure applications with minimal physical effort.
  • Constructed with durable steel and nitrile components.
  • Includes a 22 lb. steel grease bucket, steel cover, and a high-pressure hose equipped with a steel extension and coupler.
  • Features an integrated nozzle holder and rubber pads for stability.
Catalog No. 50-275-5040 Supplier Grainger Supplier No. 15F220
This high-pressure, foot-operated portable grease pump is designed for efficient, effortless lubrication of automotive, agricultural, and earth-moving equipment. The system features a design where all moving parts remain submerged in grease, which creates a wave action during operation to automatically eliminate trapped air pockets and ensure continuous lubrication without requiring a follower plate.
  • Delivers 0.07 oz. per stroke at a maximum pressure of 8,700 psi.
  • Compatible with NLGI 1 and 2 grease.
  • Operated via a convenient foot lever for high-pressure applications with minimal physical effort.
  • Constructed with durable steel and nitrile components.
  • Includes a 22 lb. steel grease bucket, steel cover, and a high-pressure hose equipped with a steel extension and coupler.
  • Features an integrated nozzle holder and rubber pads for stability.