Learn More
MSC MSC Industrial Supply Co. Grease Trap Cleaner, Ultra-Concentrated, 55 Gal.
Catalog No. 502818819
Encompass
This ultra-concentrated grease trap cleaner is designed for effective cleaning and maintenance of grease traps in various industrial and commercial settings.
  • Supplied in a 55 gal drum
  • Features ultra strength dilution at 5:1
  • Provides industrial strength dilution at 10:1
  • Offers commercial strength dilution at 16:1
Catalog No. 50-281-8819 Supplier MSC Supplier No. 08489163
This ultra-concentrated grease trap cleaner is designed for effective cleaning and maintenance of grease traps in various industrial and commercial settings.
  • Supplied in a 55 gal drum
  • Features ultra strength dilution at 5:1
  • Provides industrial strength dilution at 10:1
  • Offers commercial strength dilution at 16:1