Learn More
Wilmad Labglass GREASE VAC LVP KRYTOX 2OZ.
Supplier:  Wilmad Labglass LG9614100
Encompass
Grease, Krytox, high Vacuum Grease, LVP, Very Low Vapor Pressure, highly Inert, Nonflammable
Catalog No. 50-800-664
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.