Learn More
Grainger Crown Verity Grease Water Tray, 1 ea
Catalog No. 502794876
Encompass_Preferred
This grease water tray serves as a replacement component for outdoor grills, designed to maintain optimal equipment performance by collecting grease and water. It is engineered for compatibility with specific Crown Verity grill models to ensure a proper fit during maintenance or repair.
  • Compatible with built-in grills, gas grills, portable gas griddles, and portable gas grills
  • Designed for use with models including MCB-36, MCB-36 PKG, MCB-36 PKG NG, MG-36, and PCB-36
  • Durable construction suitable for outdoor cooking equipment requirements
Catalog No. 50-279-4876 Supplier Grainger Supplier No. 19YZ51
This grease water tray serves as a replacement component for outdoor grills, designed to maintain optimal equipment performance by collecting grease and water. It is engineered for compatibility with specific Crown Verity grill models to ensure a proper fit during maintenance or repair.
  • Compatible with built-in grills, gas grills, portable gas griddles, and portable gas grills
  • Designed for use with models including MCB-36, MCB-36 PKG, MCB-36 PKG NG, MG-36, and PCB-36
  • Durable construction suitable for outdoor cooking equipment requirements