Learn More
Takara Bio GREAT ESCAPE SEAP CHEMIL. KIT
SDP
Catalog No. NC1700054
Encompass
The Great EscAPe SEAP Chemiluminescence Kit 2.0 is a complete system for the detection of secreted alkaline phosphatase (SEAP) activity in the growth media from mammalian cells expressing this enzyme. The kit contains reagents necessary for the chemiluminescent detection of SEAP activity using a 96-well plate luminometer. The assay can also be performed in single tubes and measured in a tube luminometer. It allows you to monitor promoter activity over time, because samples of the cell media can be taken repeatedly and tested without lysing cells.
Catalog No. NC1700054 Supplier Takara Bio Supplier No. 631738
The Great EscAPe SEAP Chemiluminescence Kit 2.0 is a complete system for the detection of secreted alkaline phosphatase (SEAP) activity in the growth media from mammalian cells expressing this enzyme. The kit contains reagents necessary for the chemiluminescent detection of SEAP activity using a 96-well plate luminometer. The assay can also be performed in single tubes and measured in a tube luminometer. It allows you to monitor promoter activity over time, because samples of the cell media can be taken repeatedly and tested without lysing cells.