Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology GREB1 Monoclonal Antibody, Rabbit, Human, Unconjugated, 100 μL
SDP

Catalog No. 502732292
Encompass

This rabbit monoclonal antibody is designed to recognize endogenous levels of total GREB1 protein. The antibody is produced by immunizing animals with a synthetic peptide corresponding to residues surrounding Pro242 of human GREB1 protein. GREB1 functions as an estrogen receptor (ER) coactivator and has been implicated in various ER-based pathologies as well as androgen receptor-related signaling in prostate cancer.

  • Host species: Rabbit
  • Reactivity: Human
  • Validated applications: Western blotting (1:1,000 dilution) and immunoprecipitation (1:100 dilution)
  • Molecular weight: 216 kDa
  • Formulation: 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and <0.02% sodium azide
  • Recommended storage: -20°C (do not aliquot)

Catalog No. 50-273-2292 Supplier Cell Signaling Technology Supplier No. 65171S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This rabbit monoclonal antibody is designed to recognize endogenous levels of total GREB1 protein. The antibody is produced by immunizing animals with a synthetic peptide corresponding to residues surrounding Pro242 of human GREB1 protein. GREB1 functions as an estrogen receptor (ER) coactivator and has been implicated in various ER-based pathologies as well as androgen receptor-related signaling in prostate cancer.

  • Host species: Rabbit
  • Reactivity: Human
  • Validated applications: Western blotting (1:1,000 dilution) and immunoprecipitation (1:100 dilution)
  • Molecular weight: 216 kDa
  • Formulation: 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and <0.02% sodium azide
  • Recommended storage: -20°C (do not aliquot)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.