Learn More
Greek Waiter's Tray
SDPSCI_ED
Catalog No. s96827
Change view
Click to view available options
:
Greek waiter tray
Catalog No.
S96827 Greek waiter tray
1 options
Catalog No. S96827 Supplier Science First Supplier No. 6111820
Includes
Attached pendulum and instructions
Place a clear container of colored water on the platform of the tray and start swinging from side to side. With practice, students should be able to swing in wide circles and even upside down.
  • Durable wood tray doubles as a laboratory support stand
  • Can be clamped vertically or horizontally
  • Net force acts on container radially, toward the center
  • Container remains on the tray along with its contents
Magnets (not included) can be added to the base for use as a pendulum to demonstrate chaotic motion.
Specifications
Product Type Greek Waiter Tray
Includes Attached pendulum and instruction