Learn More
Enzo Life Sciences SEEBRIGHT Green 496 dUTP (lyophilized) (50nmol)
SDP
Catalog No. 502012546
Encompass_Preferred
Green 496 [5-Fluorescein] dUTP can replace TTP in reactions in which it serves as a substrate for E. coli DNA polymerase (holoenzyme and Klenow fragment), T4 and Taq DNA polymerases, reverse transcriptase (from AMV and M-MuLV) and terminal transferase. Fluorescently labeled probes can be prepared with this fluorescent nucleotide by a variety of methods including nick translation, random prime labeling, cDNA labeling and 3’-end labeling. Probes generated by these methods are suitable for use for the identification of specific sequences by in situ hybridization procedures on fixed cells and tissues by direct fluorescence detection. Green 496 dUTP can also be used for multicolor fluorescence labeling. This labeled dUTP can be used with the Nick Translation DNA Labeling System 2.0 (ENZ-GEN111). Alternative name: 5-Fluorescein dUTP. Quantity: Sufficient for approximately 98 reactions, following the recommended protocol of ENZ-GEN111. Purity: ≥93% (HPLC). Formulation: Lyophilized.
Catalog No. 50-201-2546 Supplier Enzo Life Sciences Supplier No. ENZ42831L0050
Green 496 [5-Fluorescein] dUTP can replace TTP in reactions in which it serves as a substrate for E. coli DNA polymerase (holoenzyme and Klenow fragment), T4 and Taq DNA polymerases, reverse transcriptase (from AMV and M-MuLV) and terminal transferase. Fluorescently labeled probes can be prepared with this fluorescent nucleotide by a variety of methods including nick translation, random prime labeling, cDNA labeling and 3’-end labeling. Probes generated by these methods are suitable for use for the identification of specific sequences by in situ hybridization procedures on fixed cells and tissues by direct fluorescence detection. Green 496 dUTP can also be used for multicolor fluorescence labeling. This labeled dUTP can be used with the Nick Translation DNA Labeling System 2.0 (ENZ-GEN111). Alternative name: 5-Fluorescein dUTP. Quantity: Sufficient for approximately 98 reactions, following the recommended protocol of ENZ-GEN111. Purity: ≥93% (HPLC). Formulation: Lyophilized.