Learn More
Perkin Elmer US LLC Green, Capillary split/splitless deactivated glass liners with deactivated wool, Pkg. 5
Supplier:  Perkin Elmer US LLC N9306233
Encompass_Preferred
Capillary splitless deactivated glass liners 4mm I.D. x 6.2mm O.D. x 92.1mm with deactivated wool
Catalog No. 50-210-4666