Learn More
Leica Biosystems GREEN CASSETTES 1000/CS
Catalog No. NC0065595
Encompass_Preferred
IP C Cassettes III are one-piece processing cassettes with a 35 degree writing surface. The cassettes are taped for easy loading into ink printers. Available in eleven colors in quantities of 1000.
Catalog No. NC0065595 Supplier Leica Biosystems Supplier No. 38440202
IP C Cassettes III are one-piece processing cassettes with a 35 degree writing surface. The cassettes are taped for easy loading into ink printers. Available in eleven colors in quantities of 1000.