Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOJO™ Green Certified Foam Hand Cleaner Refill
GREENER_CHOICESDP

Catalog No. 19043923 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Volume (Metric):
1200 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
19-043-923 1200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-043-923 Supplier GOJO™ Supplier No. 566502
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mild, readily biodegradable foam soap formula for general use.

  • Dermatologist tested to be mild
  • SANITARY SEALED™ refill helps prevent contamination
  • ECOLOGO™ Certified to Standard Name UL 2784
  • USDA Certified Biobased Product
  • Fresh dispensing valve with each refill
TRUSTED_SUSTAINABILITY

Specifications

Product Type Anti-microbial Hand Wash
Color Clear
Form Foam
For Use With (Equipment) TFX™ Dispenser
Quantity 2/Pk.
Size 1.2 L
Volume (Metric) 1200 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.