Learn More
Cayman Chemical Green CMFDA, Purity: ≥95%, CAS: 136832-63-8, 10 mg
SDP
Catalog No. 502985472
Encompass_Preferred
A cell-permeable fluorescent probe that freely enters living cells, where it is transformed into a cell-impermeable, soluble, fluorescent compound; does not affect cell viability or proliferation and stably fluoresces for at least 72 hours, allowing for its use in cell tracking
Catalog No. 50-298-5472 Supplier Cayman Chemical Supplier No. 1958310 MG
A cell-permeable fluorescent probe that freely enters living cells, where it is transformed into a cell-impermeable, soluble, fluorescent compound; does not affect cell viability or proliferation and stably fluoresces for at least 72 hours, allowing for its use in cell tracking