Learn More
AdipoGen Green CMFDA
SDP
Supplier:  AdipoGen CDXC0103M005
Encompass_Preferred
Chemical. CAS 136832-63-8. Formula C25H17ClO7. MW 464.85. Synthetic. Thiol-reactive, cell permeant green fluorescent probe with high selectivity Ex/Em 492/517nm. This fluorescent dye is well suited for monitoring cell movement or location. Freely passes through cell membranes, where it is transformed into cell membrane-impermeant reaction products and is converted to the highly fluorescent probe fluorescein once the acetate groups are cleaved by intracellular esterases. The dye exhibits ideal tracking properties it is stable, nontoxic at working concentrations, well retained in cells and brightly fluorescent at physiological pH. The green excitation/emission spectra are well separated from GFP green fluorescent protein and RFP red fluorescent protein spectra allowing for multiplexing.
Catalog No. 50-256-1470