Learn More
MOLECULAR DEPOT LLC Green Counterstain - 50 mL
Catalog No. 5000013444
Encompass_Preferred
Green Nuclear Counterstain for IHC consists of green nuclear counterstain solution containing methyl green This product has been used as molecular tool for various immunohistochemistry applications It has also been used in a wide array of other biochemical and immunological applications For in vitro research use only
Catalog No. 50-000-13444 Supplier MOLECULAR DEPOT LLC Supplier No. B2010900
Green Nuclear Counterstain for IHC consists of green nuclear counterstain solution containing methyl green This product has been used as molecular tool for various immunohistochemistry applications It has also been used in a wide array of other biochemical and immunological applications For in vitro research use only