Learn More
Andwin Scientific GREEN DISP SPATULA/PK150 310MM
SDP
Catalog No. NC0189047
Encompass_Preferred
The disposable spatulas features a unique double-ended design that allows this product to be used as a spoon, scoop, scraper, stirring rod, or policeman. This product is disposable and economical. It can be used for transferring, manipulating, and mixing materials and can also reduce the chance of cross-contamination. This product is sold in a case of 150 blue spatulas.green The V design at the neck of the disposable laboratory spatula is a trademark of LevGo, Inc
Catalog No. NC0189047 Supplier Andwin Scientific Supplier No. 80081192
The disposable spatulas features a unique double-ended design that allows this product to be used as a spoon, scoop, scraper, stirring rod, or policeman. This product is disposable and economical. It can be used for transferring, manipulating, and mixing materials and can also reduce the chance of cross-contamination. This product is sold in a case of 150 blue spatulas.green The V design at the neck of the disposable laboratory spatula is a trademark of LevGo, Inc