Learn More
Abcam Safe Image Green DNA Stain, 1ML
Catalog No. 502215690
Encompass_Preferred
The Green DNA Stain ab286890 (formerly SafeImage Green DNA Stain; M1194-1000) represents a new and safe nucleic acid stain for the visualization of double-stranded DNA (dsDNA), single-stranded DNA (ssDNA), and RNA in agarose and polyacrylamide gels.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-221-5690 Supplier Abcam Supplier No. AB2868901ML
The Green DNA Stain ab286890 (formerly SafeImage Green DNA Stain; M1194-1000) represents a new and safe nucleic acid stain for the visualization of double-stranded DNA (dsDNA), single-stranded DNA (ssDNA), and RNA in agarose and polyacrylamide gels.
The product is subject to the following: Abcam Restricted Use Statement