Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Green DND-26 | 220524-71-0 | 99.8% | 362.23 g/mol | C18H25BF2N4O | 10 MG
SDP

Catalog No. 5000296942
Encompass_Preferred

Green DND-26 is a green fluorescent lysosomal probe used for staining and imaging lysosomes in live or fixed cells. It permeates cell membranes and accumulates in acidic compartments, enabling visualization of lysosomal structure and related cellular processes.

  • Excitation/emission maxima 504/511 nm.
  • Molecular weight 362.23 g/mol.
  • Purity approximately 99.8%.
  • Physical form solid, orange to red; store solid at -20°C and protect from light.
  • In solvent: store at -80°C up to 6 months or -20°C up to 1 month, protected from light.
  • Applications suitable for fluorescence microscopy and lysosome biology studies.
  • Available in multiple pack sizes, e.g., 5-500 mg, including a 10 mg pack.

Catalog No. 50-002-96942 Supplier Medchemexpress LLC Supplier No. HYD129610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Green DND-26 is a green fluorescent lysosomal probe used for staining and imaging lysosomes in live or fixed cells. It permeates cell membranes and accumulates in acidic compartments, enabling visualization of lysosomal structure and related cellular processes.

  • Excitation/emission maxima 504/511 nm.
  • Molecular weight 362.23 g/mol.
  • Purity approximately 99.8%.
  • Physical form solid, orange to red; store solid at -20°C and protect from light.
  • In solvent: store at -80°C up to 6 months or -20°C up to 1 month, protected from light.
  • Applications suitable for fluorescence microscopy and lysosome biology studies.
  • Available in multiple pack sizes, e.g., 5-500 mg, including a 10 mg pack.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.