Learn More
MSC Betco Restroom Cleaner, Fresh Mint Scent, 32 oz, Ready-to-Use
Catalog No. 502839212
Encompass
This restroom cleaner provides professional performance for daily use, effectively removing difficult hard water stains and soap scum from various surfaces. Its formula contains organic acids that inhibit scaling and soil buildup, and non-corrosive cleaning agents to tackle tough deposits. It leaves a pleasant fresh mint fragrance, meeting Green Earth criteria.
  • Non-corrosive acid removes tough soap scum deposits.
  • Pleasant fresh mint fragrance.
  • Contains organic acids, powerful cleaning agents.
  • Inhibits scaling and soil buildup.
  • Ready to use.
  • Meets Green Earth criteria.
  • Ideal for cleaning sinks, counters, fixtures, tubs, floors, walls, glazed tile, and porcelain.
Catalog No. 50-283-9212 Supplier MSC Supplier No. 72906902
This restroom cleaner provides professional performance for daily use, effectively removing difficult hard water stains and soap scum from various surfaces. Its formula contains organic acids that inhibit scaling and soil buildup, and non-corrosive cleaning agents to tackle tough deposits. It leaves a pleasant fresh mint fragrance, meeting Green Earth criteria.
  • Non-corrosive acid removes tough soap scum deposits.
  • Pleasant fresh mint fragrance.
  • Contains organic acids, powerful cleaning agents.
  • Inhibits scaling and soil buildup.
  • Ready to use.
  • Meets Green Earth criteria.
  • Ideal for cleaning sinks, counters, fixtures, tubs, floors, walls, glazed tile, and porcelain.