Learn More
Montana Molecular Endosome targeted mNeon Green (Green EndoPaint) Expression Kit - Big Sky BacMam, 5mL - Montana Molecular
SDP
Catalog No. NC3787563
Encompass_Preferred
Bright, green fluorescent endosomal label for live-cell imaging—no fixation, staining, or cell lysis required. Visualize endosomal morphology and spatial dynamics in real time over hours to days in living cells. Delivered using Montana Molecular’s Big Sky BacMam vector for robust expression across a wide range of cell types, including primary cultures and iPSCs. Endosomal targeting is achieved via the FYVE domain. Ideal for dynamic, non-destructive studies of cellular phenotypes, endosomal organization, and high-content imaging.
Catalog No. NC3787563 Supplier Montana Molecular Supplier No. P1080G005
Bright, green fluorescent endosomal label for live-cell imaging—no fixation, staining, or cell lysis required. Visualize endosomal morphology and spatial dynamics in real time over hours to days in living cells. Delivered using Montana Molecular’s Big Sky BacMam vector for robust expression across a wide range of cell types, including primary cultures and iPSCs. Endosomal targeting is achieved via the FYVE domain. Ideal for dynamic, non-destructive studies of cellular phenotypes, endosomal organization, and high-content imaging.