Learn More
Sigma Aldrich Fine Chemicals Biosciences PKH67 Green Fluorescent Cell Linker Kit for General Cell Membrane Labeling, 1 kit
Catalog No. NC0949883
Encompass
PKH67 Green Fluorescent Cell Linker Kit for General Cell Membrane Labeling has been used: To label and further investigate the exosomes expelled from cells in triple-negative breast cancer condition, to label apoptotic cells, in order to study the role of alphaνbeta5 receptor in both binding and internalization of apoptotic cells, and in fluorescence imaging.
Catalog No. NC0949883 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. PKH67GL1KT
PKH67 Green Fluorescent Cell Linker Kit for General Cell Membrane Labeling has been used: To label and further investigate the exosomes expelled from cells in triple-negative breast cancer condition, to label apoptotic cells, in order to study the role of alphaνbeta5 receptor in both binding and internalization of apoptotic cells, and in fluorescence imaging.